Läs mer
1-(2-Chloro-4-(4-chlorophenoxy)phenyl)-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol, TRC
Produktkod. 30350216
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350216 5 mg
30357082 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4292535MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 117018-19-6
Molekylformel C16 H13 Cl2 N3 O2
Molekylvikt (g/mol) 350.2
Synonym α-[2-Chloro-4-(4-chlorophenoxy)phenyl]-1H-1,2,4-triazole-1-ethanol, CGA 205375
IUPAC-namn 1-[2-chloro-4-(4-chlorophenoxy)phenyl]-2-(1,2,4-triazol-1-yl)ethanol
LEDER OC(Cn1cncn1)c2ccc(Oc3ccc(Cl)cc3)cc2Cl
Specifikationer
Kemiskt namn eller material Difenoconazole-alcohol
Kvantitet 5 mg
InChI formel InChI=1S/C16H13Cl2N3O2/c17-11-1-3-12(4-2-11)23-13-5-6-14(15(18)7-13)16(22)8-21-10-19-9-20-21/h1-7,9-10,16,22H,8H2
Formel vikt 349.0385