missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloro-4-nitrophenyl)azepane, 95%, Thermo Scientific™

Catalog Number p-7054774
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1260897-00-4
C12H15ClN2O2
254.714
YQJCKUFYSNESGZ-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl azepane
100913297
1-(2-chloro-4-nitrophenyl)azepane
C1CCCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
This item is not returnable. View return policy
1260897-00-4
C12H15ClN2O2
254.714
YQJCKUFYSNESGZ-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl azepane
100913297
1-(2-chloro-4-nitrophenyl)azepane
C1CCCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Formula Weight 254.71
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)azepane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'