missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H63094.03

Additional Details : CAS Number : 59504-30-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499588

  • 1002.82 SEK / 1g


Estimated Shipment date
from Supplier 24-07-2019
Add to basket

Specifications

Chemical Identifiers

59504-30-2
226.66
1-2-chloro-6-nitrophenyl pyrrolidine, 6-chloro-2-nitrophenyl pyrrolidine, 2-pyrrolidin-1-yl-3-chloro-1-nitrobenzene
1-(2-chloro-6-nitrophenyl)pyrrolidine
C10H11ClN2O2
SHWAUCHKWGIYJA-UHFFFAOYSA-N
5112761
C1CCN(C1)C2=C(C=CC=C2Cl)[N+](=O)[O-]

Specifications

1-(2-Chloro-6-nitrophenyl)pyrrolidine
59504-30-2
4317456
SHWAUCHKWGIYJA-UHFFFAOYSA-N
1-(2-chloro-6-nitrophenyl)pyrrolidine
5112761
Solid
1g
C10H11ClN2O2
1-2-chloro-6-nitrophenyl pyrrolidine, 6-chloro-2-nitrophenyl pyrrolidine, 2-pyrrolidin-1-yl-3-chloro-1-nitrobenzene
C1CCN(C1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
226.66
226.66
97%
Videos
SDS