Läs mer
1-(2-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic Acid Methyl Ester, TRC
Produktkod. 30350410
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30350410 10 mg
1 options
Produktkod. 30350410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36440510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(2-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic Acid Methyl Ester
CAS 1568-35-0
Molekylformel C20H18ClNO4
Kvantitet 10 mg
Synonym 1-(o-Chlorobenzoyl)-5-methoxy-2-methyl-indole-3-acetic Acid Methyl Ester
InChI formel InChI=1S/C20H18ClNO4/c1-12-15(11-19(23)26-3)16-10-13(25-2)8-9-18(16)22(12)20(24)14-6-4-5-7-17(14)21/h4-10H,11H2,1-3H3
LEDER O=C(OC)CC1=C(C)N(C(C2=C(Cl)C=CC=C2)=O)C3=CC=C(OC)C=C31
IUPAC-namn methyl 2-(1-(2-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetate
Molekylvikt (g/mol) 371.81
Formel vikt 371.09