missing translation for 'onlineSavingsMsg'
Läs mer

1-[(2-Klorofenyl)difenylmetyl]-1H-pyrazol, Mikromol™

Product Code. 30051745
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30051745 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051745 Supplier Mikromol Supplier No. MM0015.090025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol – din pålitliga källa för högkvalitativa läkemedelsreferensstandarder. Stöder noggrann, regelrätt analys med material som är ackrediterat 17034 ISO och utformat för att säkerställa förtroende för varje resultat.

1-[(2-Klorofenyl)difenylmetyl]-1H-pyrazol
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
CAS 289905-88-0
Analyt- eller komponentnamn 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
Frakt skick Rumstemperatur
Molekylformel C22 H17 Cl N2
Certifikat och efterlevnad ISO 9001
Synonym 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
InChI formel InChI=1S/C22H17ClN2/c23-21-15-8-7-14-20(21)22(25-17-9-16-24-25,18-10-3-1-4-11-18)19-12-5-2-6-13-19/h1-17H
LEDER Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4cccn4
IUPAC-namn 1-[(2-klorofenyl)-difenylmetyl] pyrazol
Molekylvikt (g/mol) 344.84
Kvantitet 25 mg
Formel vikt 344,108 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.