Läs mer
1,2-Cycloheptanedione, TRC
Produktkod. 30368349
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30368349 10 mg
1 options
Product Code. 30368349 Supplier Toronto Research Chemicals Supplier No. TRCC98725510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1,2-Cycloheptanedione
CAS 3008-39-7
Molekylformel C7H10O2
Kvantitet 10 mg
InChI formel InChI=1S/C7H10O2/c8-6-4-2-1-3-5-7(6)9/h1-5H2
LEDER O=C1CCCCCC1=O
IUPAC-namn cycloheptane-1,2-dione
Molekylvikt (g/mol) 126.15
Formel vikt 126.0681