Läs mer
1,2-Cyclohexanedicarboxylic Acid Mono 4-Methyl-7-carboxy-heptyl Ester, TRC
Produktkod. 17971495
Change view
Kvantitet:
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17971495 10 mg -
17964715 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17971495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98731510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1637562-51-6
Molekylformel C17H28O6
Molekylvikt (g/mol) 328.4
Synonym 2-(((7-Carboxy-4-methylheptyl)oxy)carbonyl)cyclohexanecarboxylic Acid, CX-MINCH
IUPAC-namn 2-(7-carboxy-4-methylheptoxy)carbonylcyclohexane-1-carboxylic acid
LEDER O=C(O)C1C(C(OCCCC(C)CCCC(O)=O)=O)CCCC1
Specifikationer
Kemiskt namn eller material 1,2-Cyclohexanedicarboxylic Acid Mono 4-Methyl-7-carboxy-heptyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C17H28O6/c1-12(6-4-10-15(18)19)7-5-11-23-17(22)14-9-3-2-8-13(14)16(20)21/h12-14H,2-11H2,1H3,(H,18,19)(H,20,21)
Formel vikt 328.1886
Procent renhet >95%