missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexanedione dioxime, 97%, Thermo Scientific Chemicals
625.64 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.
Solubility
Soluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.
Solubility
Soluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 181°C to 188°C (decomposition) |
Quantity | 5 g |
Beilstein | 1909047 |
Merck Index | 14,6560 |
Solubility Information | Soluble in water. |
Formula Weight | 142.16 |
Percent Purity | 97% |
Chemical Name or Material | 1,2-Cyclohexanedione dioxime |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'