Läs mer
1,2-Dehydro-3-oxo Rocuronium Bromide, TRC
Produktkod. 30136794
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136794 50 mg
30007291 1 mg
30003380 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136794 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23051550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1190105-67-9
Molekylformel C32 H49 N2 O4 . Br
Molekylvikt (g/mol) 605.65
Synonym Pyrrolidinium, 1-[(5α,16β,17β)-17-(acetyloxy)-2-(4-morpholinyl)-3-oxoandrost-1-en-16-yl]-1-(2-propen-1-yl)-, bromide (1:1), 1-[17β-Acetoxy-2-(morpholin-4-yl)-3-oxo-5α-androst-1-en-16β-yl]-1-(prop-2-enyl)pyrrolidinium Bromide
IUPAC-namn [(5S,8R,9S,10S,13S,14S,16S,17R)-10,13-dimethyl-2-morpholin-4-yl-3-oxo-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate;bromide
LEDER [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4CC(=O)C(=C[C@]4(C)[C@H]3CC[C@]12C)N5CCOCC5)[N+]6(CC=C)CCCC6
Specifikationer
Kemiskt namn eller material 1,2-dehydro-3-oxo Rocuronium bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C32H49N2O4.BrH/c1-5-14-34(15-6-7-16-34)28-20-26-24-9-8-23-19-29(36)27(33-12-17-37-18-13-33)21-32(23,4)25(24)10-11-31(26,3)30(28)38-22(2)35;/h5,21,23-26,28,30H,1,6-20H2,2-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,28-,30-,31-,32-;/m0./s1
Formel vikt 604.2876
Procent renhet >95%