missing translation for 'onlineSavingsMsg'
Läs mer

1-(2'-Deoxy-5'-O-DMT-3'-O-nitrophenylsulphonyl-b-D-lyxofuranosyl)thymine, TRC

Produktkod. 30360474
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360474 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30360474 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD210870250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1-(2'-Deoxy-5'-O-DMT-3'-O-nitrophenylsulphonyl-b-D-lyxofuranosyl)thymine
CAS 444717-20-8
Molekylformel C37H35N3O11S
Kvantitet 250 mg
Synonym 1-[5-O-[Bis(4-methoxyphenyl)phenylmethyl]-2-deoxy-3-O-[(4-nitrophenyl)sulfonyl]-beta-D-threo-pentofuranosyl]-5-methyl-2,4(1H,3H)-pyrimidinedione
InChI formel InChI=1S/C37H35N3O11S/c1-24-22-39(36(42)38-35(24)41)34-21-31(51-52(45,46)33-12-8-7-11-30(33)40(43)44)32(50-34)23-49-37(25-9-5-4-6-10-25,26-13-17-28(47-2)18-14-26)27-15-19-29(48-3)20-16-27/h4-20,22,31-32,34H,21,23H2,1-3H3,(H,38,41,42)/t31-,32-,34-/m1/s1
LEDER CC1=CN([C@H]2C[C@@H](OS(=O)(C3=CC=CC=C3[N+]([O-])=O)=O)[C@@H](COC(C4=CC=C(OC)C=C4)(C5=CC=C(OC)C=C5)C6=CC=CC=C6)O2)C(NC1=O)=O
IUPAC-namn (2R,3R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl 2-nitrobenzenesulfonate
Molekylvikt (g/mol) 729.75
Formel vikt 729.2
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.