Läs mer
1,2-Diamino-4,5-ethylenedioxybenzene, Dihydrochloride, TRC
Produktkod. 30354020
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30354020 10 mg
1 options
Produktkod. 30354020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41632010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2-Diamino-4,5-ethylenedioxybenzene, Dihydrochloride
CAS 73448-02-9
Molekylformel C8 H10 N2 O2
Kvantitet 10 mg
Synonym 1,4-Benzodioxin-6,7-diamine, 2,3-dihydro-, hydrochloride (1:2), 1,4-Benzodioxin-6,7-diamine, 2,3-dihydro-, dihydrochloride (9CI), 2,3-Dihydro-1,4-benzodioxin-6,7-diamine dihydrochloride, 1,2-Diamino-4,5-ethylenedioxybenzene dihydrochloride, 2,3-Dihydrobenzo[1,4]dioxine-6,7-diamine dihydrochloride
InChI formel InChI=1S/C8H10N2O2/c9-5-3-7-8(4-6(5)10)12-2-1-11-7/h3-4H,1-2,9-10H2
LEDER Nc1cc2OCCOc2cc1N
IUPAC-namn 2,3-dihydro-1,4-benzodioxine-6,7-diamine
Molekylvikt (g/mol) 166.18
Formel vikt 166.0742