missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexane, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-100009
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
694-83-7
C6H14N2
114.19
MFCD00001491
SSJXIUAHEKJCMH-UHFFFAOYSA-N
1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
4610
cyclohexane-1,2-diamine
C1CCC(C(C1)N)N
This item is not returnable. View return policy
694-83-7
C6H14N2
114.19
MFCD00001491
SSJXIUAHEKJCMH-UHFFFAOYSA-N
1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
4610
cyclohexane-1,2-diamine
C1CCC(C(C1)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.0°C to 15.0°C
Density 0.9300g/mL
Boiling Point 188.0°C to 192.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NH2)2
Refractive Index 1.4860 to 1.4920
Quantity 250 g
Beilstein 13, 1
Specific Gravity 0.93
Solubility Information Solubility in water: miscible.
Viscosity 7.2 mPa.s (21°C)
Formula Weight 114.19
Percent Purity 99%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 2-Diaminocyclohexane, tech., 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'