Learn More
1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals
Catalog Number p-7022135
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
583-53-9
C6H4Br2
235.906
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br
583-53-9
C6H4Br2
235.906
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 4°C to 6°C
Density 1.985
Boiling Point 224°C to 226°C
Flash Point 91°C (195°F)
Odor Strong
Refractive Index 1.611
Quantity 25 g
UN Number UN3082
Beilstein 970241
Solubility Information Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobenzene