missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromopropane, 98%, Thermo Scientific Chemicals

Catalog Number p-7026780
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
78-75-1
C3H6Br2
201.889
MFCD00000148
XFNJYAKDBJUJAJ-UHFFFAOYSA-N
propylene dibromide, propane, 1,2-dibromo, propylene bromide, 2,3-dibromopropane, ccris 6710, ch2brchbrch3, propane, 1,2-dibromo-, r, propane, 1,2-dibromo-, s, 1,2 dibromopropane, 1,2-dibromo-propane
6553
1,2-dibromopropane
CC(CBr)Br
This item is not returnable. View return policy
78-75-1
C3H6Br2
201.889
MFCD00000148
XFNJYAKDBJUJAJ-UHFFFAOYSA-N
propylene dibromide, propane, 1,2-dibromo, propylene bromide, 2,3-dibromopropane, ccris 6710, ch2brchbrch3, propane, 1,2-dibromo-, r, propane, 1,2-dibromo-, s, 1,2 dibromopropane, 1,2-dibromo-propane
6553
1,2-dibromopropane
CC(CBr)Br

1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

Solubility
Soluble in water (slightly), ether, and alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -55°C
Density 1.932
Boiling Point 140°C to 142°C
Flash Point 35°C (95°F)
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 1718884
Merck Index 14,7853
Solubility Information Soluble in water (slightly),ether,and alcohol.
Formula Weight 201.9
Percent Purity 98%
Chemical Name or Material 1,2-Dibromopropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'