missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%, Reagent Grade, Honeywell™

Catalog Number p-7120012
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -17°C
Color Colorless to Yellow
Density 1.306g/mL at 25°C
Boiling Point 180°C (1013 hPa)
Explosion Limit 0.092
Flash Point 66°C (150.8°F)
Ignition Point 1198°F
Odor Aromatic
Packaging Glass bottle
Quantity 100 mL
UN Number UN1591
Beilstein 606078
Vapor Density 5.1 (vs air)
Formula Weight 147g/mol
Vapor Pressure 1.6mmHg (35°C), 1.2mmHg (20°C)
Percent Purity 0.99
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'