Läs mer
1,2-Dihydro-1,1,6-trimethylnaphthalene (>80%), TRC
Produktkod. 17929844
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17929844 100 mg
17998304 25 mg
17918424 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17929844 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453440100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30364-38-6
Molekylformel C13 H16
Molekylvikt (g/mol) 172.27
Synonym 1,1,6-Trimethyl-1,2-dihydronaphthalene, 1,2-Dihydro-1,1,6-trimethylnaphthalene, Dehydro-ar-ionene
IUPAC-namn 1,1,6-trimethyl-2H-naphthalene
LEDER Cc1ccc2c(C=CCC2(C)C)c1
Specifikationer
Kemiskt namn eller material 1,2-Dihydro-1,1,6-trimethylnaphthalene
Kvantitet 100 mg
InChI formel InChI=1S/C13H16/c1-10-6-7-12-11(9-10)5-4-8-13(12,2)3/h4-7,9H,8H2,1-3H3
Formel vikt 172.1252