Läs mer
1,2-Dihydro Desoxymetasone, TRC
Produktkod. 30378797
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378797 25 mg
30370548 10 mg
30370709 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378797 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44887525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 432-54-2
Molekylformel C22 H31 F O4
Molekylvikt (g/mol) 378.48
Synonym Pregn-4-ene-3,20-dione, 9-fluoro-11,21-dihydroxy-16-methyl-, (11β,16α)- (9CI), Pregn-4-ene-3,20-dione, 9-fluoro-11β,21-dihydroxy-16α-methyl- (8CI), Pregn-4-ene-3,20-dione, 9α-fluoro-11β,21-dihydroxy-16α-methyl- (7CI), 9-Fluoro-11β,21-dihydroxy-16α-methylpregna-4-ene-3,20-dione, 1,2-Dihydro Desoxymetasone, 1,2-Dihydrodesoxymetasone
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-2,6,7,8,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@H]1C(=O)CO
Specifikationer
Kemiskt namn eller material 1,2-Dihydro Desoxymetasone
Kvantitet 25 mg
InChI formel InChI=1S/C22H31FO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,23)18(27)10-20(16,2)19(12)17(26)11-24/h9,12,15-16,18-19,24,27H,4-8,10-11H2,1-3H3/t12-,15+,16+,18+,19-,20+,21+,22+/m1/s1
Formel vikt 378.2206