Läs mer
1,2-Dihydro Dexamethasone, TRC
Produktkod. 30008201
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008201 50 mg
30005904 25 mg
30011070 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008201 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44901050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 426-17-5
Molekylformel C22 H31 F O5
Molekylvikt (g/mol) 394.48
Synonym 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregn-4-ene-3,20-dione, Dexamethasone Imp. C (EP), Pregn-4-ene-3,20-dione, 9-fluoro-11,17,21-trihydroxy-16-methyl-, (11β,16α)-, Pregn-4-ene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl- (7CI,8CI), (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregn-4-ene-3,20-dione, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregn-4-ene-3,20-dione, RU 25008
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregn-4-ene-3,20-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H31FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h9,12,15-17,24,26,28H,4-8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formel vikt 394.2156
Procent renhet >95%