Läs mer
1,2-Dihydrobudesonide (Mixture of diastereomers), TRC
Produktkod. 30136795
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136795 250 mg
30009336 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229330250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137174-25-5
Molekylformel C25H36O6
Molekylvikt (g/mol) 432.55
Synonym Budesonide Imp. G (EP), 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregn-4-ene-3,20-dione, (11beta,16alpha)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxy-pregn-4-ene-3,20-dione, 1,2-Dihydrobudesonide
IUPAC-namn 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregn-4-ene-3,20-dione
LEDER CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 1,2-Dihydrobudesonide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C25H36O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h10,16-18,20-22,26,28H,4-9,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vikt 432.2512
Procent renhet >95%