Läs mer
1,2-Dihydrodesonide, TRC
Produktkod. 17974135
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974135 25 mg
17902725 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17974135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44817025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 638-93-7
Molekylformel C24 H34 O6
Molekylvikt (g/mol) 418.52
Synonym Dihydrodesonide, 11β,21-Dihydroxy-16α,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, Pregn-4-ene-3,20-dione, 11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)-, Corticosterone, 16α,17-dihydroxy-, cyclic 16,17-acetal with acetone (6CI), Pregn-4-ene-3,20-dione, 11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI), (11β,16α)-11,21-Dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, 11β,21-Dihydroxy-16α,17-isopropylidenedioxy-4-pregnene-3,20-dione, 16α-Hydroxyhydrocortisone 16,17-acetonide
LEDER CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 1,2-Dihydrodesonide
Kvantitet 25 mg
InChI formel InChI=1S/C24H34O6/c1-21(2)29-19-10-16-15-6-5-13-9-14(26)7-8-22(13,3)20(15)17(27)11-23(16,4)24(19,30-21)18(28)12-25/h9,15-17,19-20,25,27H,5-8,10-12H2,1-4H3/t15-,16-,17-,19+,20+,22-,23-,24+/m0/s1
Formel vikt 418.2355