Läs mer
1,2-Dihydroxynaphthalene, TRC
Produktkod. 30374454
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374454 50 mg
30386695 500 mg
30379937 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30374454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45416850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 574-00-5
Molekylformel C10H8O2
Molekylvikt (g/mol) 160.17
Synonym 1,2-Naphthalenediol, 1,2-Naphthoquinol, 1-Hydroxy-2-naphthol, NSC 401609
LEDER Oc1ccc2ccccc2c1O
Specifikationer
Kemiskt namn eller material 1,2-Dihydroxynaphthalene
Kvantitet 50 mg
InChI formel InChI=1S/C10H8O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,11-12H
Formel vikt 160.0524