missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt, TRC
Specifications
Specifications
| Kemiskt namn eller material | 1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt |
| CAS | 80724-31-8 |
| Molekylformel | C31H60NaO8P |
| Kvantitet | 1 mg |
| Synonym | 1,1'-[(1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl] Ester Tetradecanoic Acid Sodium Salt, (1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl Ester Tetradecanoic Acid Monosodium Salt, 1,2-Dimyristoyl-sn-glycero-3-phosphate Monosodium Salt, Monosodium 1,2-Dimyristoyl-sn-glycerophosphate |
| InChI formel | InChI=1S/C31H61O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(32)37-27-29(28-38-40(34,35)36)39-31(33)26-24-22-20-18-16-14-12-10-8-6-4-2;/h29H,3-28H2,1-2H3,(H2,34,35,36);/q;+1/p-1/t29-;/m1./s1 |
| LEDER | [Na+].CCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)[O-])OC(=O)CCCCCCCCCCCCC |
| Molekylvikt (g/mol) | 614.77 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?