Läs mer
1,2-Dioleoyloxy-3-(dimethylamino)propane, TRC
Produktkod. 30134689
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134689 1 g
30007354 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4830001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127512-29-2
Molekylformel C41 H77 N O4
Molekylvikt (g/mol) 648.05
IUPAC-namn [3-(dimethylamino)-2-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)OCC(CN(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC
Specifikationer
Kemiskt namn eller material 1,2-Dioleoyloxy-3-(dimethylamino)propane
Kvantitet 1 g
InChI formel InChI=1S/C41H77NO4/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-40(43)45-38-39(37-42(3)4)46-41(44)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h19-22,39H,5-18,23-38H2,1-4H3/b21-19-,22-20-
Formel vikt 647.5853