Läs mer
1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, TRC
Produktkod. 30372441
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30372441 250 mg
1 options
Produktkod. 30372441 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD525495250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
CAS 200880-41-7
Molekylformel C38H74NaO10P
Kvantitet 250 mg
Synonym 1,2-Dihexadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol dipalmitoyl, DPPG-Na, Hexadecanoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester
InChI formel InChI=1S/C38H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(41)45-33-36(34-47-49(43,44)46-32-35(40)31-39)48-38(42)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35-36,39-40H,3-34H2,1-2H3,(H,43,44);/q;+1/p-1/t35?,36-;/m1./s1
LEDER [Na+].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC(O)CO)OC(=O)CCCCCCCCCCCCCCC
IUPAC-namn sodium;[(2R)-2,3-di(hexadecanoyloxy)propyl] 2,3-dihydroxypropyl phosphate
Molekylvikt (g/mol) 744.95
Formel vikt 744.4917