Läs mer
1,2-Distearoyl-3-oleoyl-rac-glycerol, TRC
Produktkod. 30007734
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007734 500 mg
30005681 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD493520500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2190-29-6
Molekylformel C57 H108 O6
Molekylvikt (g/mol) 889.46
Synonym (Z)-9-Octadecenoic Acid 2,3-Bis[(1-oxooctadecyl)oxy]propyl Ester, 2,3-Distearo-1-olein, 3-Oleo-1,2-di-stearin, (±)-Glycerol 1,2-Distearate 3-oleate, 1,2-Distearo-3-olein, 1,2-Distearoyl-3-oleoyl-rac-glycerol, 1-Oleo-2,3-distearin, 1-Oleoyl distearin, 3-Oleo-1,2-distearin, Glyceryl 1-oleate-2,3-distearate
IUPAC-namn [2-octadecanoyloxy-3-[(Z)-octadec-9-enoyl]oxy-propyl] octadecanoate
LEDER CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material 1,2-Distearoyl-3-oleoyl-rac-glycerol
Kvantitet 500 mg
InChI formel InChI=1S/C57H108O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25,28,54H,4-24,26-27,29-53H2,1-3H3/b28-25-
Formel vikt 888.8146