missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Ethoxyethyl)piperazine, 97%

Brand:  Alfa Aesar™ H55186.03

Additional Details : CAS Number : 13484-38-3 Weight : 0.00100kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15461348

  • 632.48 SEK / 1g


Estimated Shipment date
from Supplier 21-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Ethoxyethyl)piperazine
13484-38-3
158.24
MFCD01320899

Specifications

1-(2-Ethoxyethyl)piperazine
224° to 225°C
1g
1.4690
97%
MFCD01320899
0.941g/mL
Colorless to dark yellow
13484-38-3
Liquid
158.24
Videos
SDS