missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Iodobenzyl)-4-methylpiperazine, ≥95%, Thermo Scientific™

Catalog Number 10416613
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 156333-97-0
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C12H17IN2
MDL Number MFCD06797860
Quantity 1 g
Synonym 1-2-iodobenzyl-4-methylpiperazine, 1-2-iodophenyl methyl-4-methylpiperazine, piperazine,1-2-iodophenyl methyl-4-methyl
InChI Key HSNIZBDTYYAXLY-UHFFFAOYSA-N
SMILES CN1CCN(CC1)CC2=CC=CC=C2I
IUPAC Name 1-[(2-iodophenyl)methyl]-4-methylpiperazine
Molecular Weight (g/mol) 316.186
PubChem CID 15643052
Formula Weight 316.18
Percent Purity 90%
Chemical Name or Material 1-(2-Iodobenzyl)-4-methylpiperazine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'