Läs mer
1-(2-Methylbenzyl)piperazine, 97%, Thermo Scientific™
Produktkod. 15461368
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15461368 5 g 5g
15451368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15461368 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H55203.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5321-47-1
Molekylformel C12H18N2
Molekylvikt (g/mol) 190.29
MDL-nummer MFCD02256026
InChI-nyckel GRVNOUKHAZXMJK-UHFFFAOYSA-N
Synonym 1-2-methylbenzyl piperazine, 1-2-methylphenyl methyl piperazine, 1-2-methylbenzyl-piperazine, 1-2-methyl-benzyl-piperazine, piperazine, 1-2-methylphenyl methyl, 2-methylphenyl methyl piperazine, 2-methylbenzylpiperazine, acmc-1asr0, 1-2-methyl-benzyl piperazine
PubChem CID 79212
IUPAC-namn 1-[(2-methylphenyl)methyl]piperazine
LEDER CC1=CC=CC=C1CN1CCNCC1
Specifikationer
Kemiskt namn eller material 1-(2-Methylbenzyl)piperazine
Densitet 0.988 g/mL
Kokpunkt 77°C to 80°C (0.1 mmHg)
Brytningsindex 1.543
Kvantitet 5 g
Formel vikt 190.29
Procent renhet 97%
Fysisk form Liquid