Läs mer
1-(2-Nitroethyl)-2-naphthol, TRC
Produktkod. 30474646
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474646 5 g
30472391 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474646 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4958955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96853-41-7
Molekylformel C12 H11NO3
Molekylvikt (g/mol) 217.22
IUPAC-namn 1-(2-nitroethyl)naphthalen-2-ol
LEDER Oc1ccc2ccccc2c1CC[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 1-(2-Nitroethyl)-2-naphthol
Kvantitet 5 g
InChI formel InChI=1S/C12H11NO3/c14-12-6-5-9-3-1-2-4-10(9)11(12)7-8-13(15)16/h1-6,14H,7-8H2
Formel vikt 217.0739