Läs mer
1,2-O-Cyclohexylidene myo-Inositol, TRC
Produktkod. 30353816
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353816 10 g
30349949 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353816 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98800010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6763-47-9
Molekylformel C12 H20 O6
Molekylvikt (g/mol) 260.28
IUPAC-namn (3aS,4S,5R,6R,7S,7aR)-spiro[3a,4,5,6,7,7a-hexahydro-1,3-benzodioxole-2,1'-cyclohexane]-4,5,6,7-tetrol
LEDER O[C@H]1[C@@]2([H])OC3(CCCCC3)O[C@@]2([H])[C@@H]([C@H]([C@@H]1O)O)O
Specifikationer
Kemiskt namn eller material 1,2-O-Cyclohexylidene-myo-inositol
Kvantitet 10 g
InChI formel InChI=1S/C12H20O6/c13-6-7(14)9(16)11-10(8(6)15)17-12(18-11)4-2-1-3-5-12/h6-11,13-16H,1-5H2/t6-,7-,8+,9+,10-,11+/m1/s1
Formel vikt 260.126