Läs mer
1,2’-O-Dimethylguanosine, TRC
Produktkod. 30134434
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134434 25 mg -
30138355 10 mg -
30003993 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46595025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73667-71-7
Molekylformel C12 H17 N5 O5
Molekylvikt (g/mol) 311.294
Synonym 1-Methyl-2'-O-methylguanosine, 2'-O-Methyl-1-methylguanosine
IUPAC-namn 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-1-methylpurin-6-one
LEDER CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1n2cnc3C(=O)N(C)C(=Nc23)N
Specifikationer
Kemiskt namn eller material 1,2'-O-Dimethylguanosine
Kvantitet 25 mg
InChI formel InChI=1S/C12H17N5O5/c1-16-10(20)6-9(15-12(16)13)17(4-14-6)11-8(21-2)7(19)5(3-18)22-11/h4-5,7-8,11,18-19H,3H2,1-2H3,(H2,13,15)/t5-,7-,8-,11-/m1/s1
Formel vikt 311.123