Learn More
1,2-Propylene Oxide, Sigma-Aldrich™
Product Code. 15859390
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859390 1 L 1 L
15849390 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15859390 Supplier Sigma Aldrich Supplier No. 8070271000
 Restricted Item
Beräknad leverans: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-56-9
Grade Research
Molecular Weight (g/mol) 58.08
Specifications
Melting Point -112°C
Density 0.83 g/cm3 at 20°C
MDL Number MFCD00005126
Quantity 1 L
Synonym 1,2-Propylene oxide, 1,2-Epoxypropane, Methyloxirane, 1,2-Propylene oxide