missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7055434
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1141878-45-6
C9H12BF3N2O3
264.011
MFCD17214248
IIRVABFYRULQMI-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydropyran-2-yl-3-trifluoromethyl pyrazole-5-boronic acid, 1-oxan-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 2-tetrahydropyran-2-yl-5-trifluoromethyl pyrazol-3-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronicacid, 2-oxan-2-yl-5-trifluoromethyl pyrazol-3-ylboronic acid, acmc-2099kl, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, boronic acid, b-1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl
53216481
[2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
B(C1=CC(=NN1C2CCCCO2)C(F)(F)F)(O)O
This item is not returnable. View return policy
1141878-45-6
C9H12BF3N2O3
264.011
MFCD17214248
IIRVABFYRULQMI-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydropyran-2-yl-3-trifluoromethyl pyrazole-5-boronic acid, 1-oxan-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 2-tetrahydropyran-2-yl-5-trifluoromethyl pyrazol-3-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronicacid, 2-oxan-2-yl-5-trifluoromethyl pyrazol-3-ylboronic acid, acmc-2099kl, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, boronic acid, b-1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl
53216481
[2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
B(C1=CC(=NN1C2CCCCO2)C(F)(F)F)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 264.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'