Läs mer
Invitrogen™ 1,25 Dihydroxy Vitamin D3 Competitive ELISA Kit
Beskrivning
The 1,25 Dihydroxyvitamin D3 (DHVD3) ELISA quantitates DHVD3 in serum, plasma and other biological fluids. Principle of the method The Competitive mulispecies ELISA research use only kit is designed to quantitatively measure 1,25 Dihydroxyvitamin D3independent of species . A 1,25 Dihydroxyvitamin D3 standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve.Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to 1,25 Dihydroxyvitamin D3 is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of 1,25 Dihydroxyvitamin D3 in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Specifikationer
Specifikationer
| Analysintervall | 7.81-500 pg/mL |
| Analyskänslighet | 4.69 pg/mL |
| Konjugera | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Produkttyp | ELISA |
| Provtyp | Cell Lysates, Plasma, Serum, Tissue Homogenates |
| Målarter | All Species |
| För användning med (utrustning) | Absorbance Microplate Reader |
| Gene Alias | CalcitriolCalcitriol; |
| Interassay CV | <10% |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.