Läs mer
1-[[(2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiranyl]methyl]-1H-1,2,4-triazole, TRC
Produktkod. 30370624
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370624 5 g
30376503 500 mg
30371107 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30370624 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4478155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127000-90-2
Molekylformel C12H11F2N3O
Molekylvikt (g/mol) 251.23
Synonym (2R,3S)-2-(2,4-Difluorophenyl)-3-methyl-[(1H-1,2,4-triazol-1-yl)methyl]oxirane, (2R,3S)-2-(2,4-Difluorophenyl)-3-methyl-2-(1H-1,2,4-triazol-1-yl)methyloxirane
LEDER C[C@@H]1O[C@@]1(Cn2cncn2)c3ccc(F)cc3F
Specifikationer
Kemiskt namn eller material 1-(((2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiranyl)methyl)-1H-1,2,4-triazole
Kvantitet 5 g
InChI formel InChI=1S/C12H11F2N3O/c1-8-12(18-8,5-17-7-15-6-16-17)10-3-2-9(13)4-11(10)14/h2-4,6-8H,5H2,1H3/t8-,12+/m0/s1
Formel vikt 251.087