Läs mer
1-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-4-(2-phenoxyethyl)-1,2,4-triazolidine-3,5-dione, TRC
Produktkod. 30361957
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361957 50 mg
30361792 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361957 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38036550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 153707-88-1
Molekylformel C23H28ClN5O3
Molekylvikt (g/mol) 457.95
Synonym BMS 180492
IUPAC-namn 1-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-4-(2-phenoxyethyl)-1,2,4-triazolidine-3,5-dione
LEDER O=C1N(CCOC2=CC=CC=C2)C(NN1CCCN3CCN(C4=CC=CC(Cl)=C4)CC3)=O
Specifikationer
Kemiskt namn eller material 1-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-4-(2-phenoxyethyl)-1,2,4-triazolidine-3,5-dione
Kvantitet 50 mg
InChI formel InChI=1S/C23H28ClN5O3/c24-19-6-4-7-20(18-19)27-14-12-26(13-15-27)10-5-11-29-23(31)28(22(30)25-29)16-17-32-21-8-2-1-3-9-21/h1-4,6-9,18H,5,10-17H2,(H,25,30)