missing translation for 'onlineSavingsMsg'
Learn More

1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 97%, Thermo Scientific™

Catalog Number p-4510525
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
852227-97-5
C17H26BNO2
287.21
MFCD07368528
IIUXWPXCCXVCPD-UHFFFAOYSA-N
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidinophenylboronic acid, pinacol ester, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidin-1-yl benzeneboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
4961251
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCCCC1
This item is not returnable. View return policy
852227-97-5
C17H26BNO2
287.21
MFCD07368528
IIUXWPXCCXVCPD-UHFFFAOYSA-N
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidinophenylboronic acid, pinacol ester, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidin-1-yl benzeneboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
4961251
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCCCC1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 287.21
Percent Purity 97%
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'