Läs mer
1-(3,4-Dichlorophenyl)-1,2-dihydronaphthalene, TRC
Produktkod. 30355310
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355310 100 mg
30353197 500 mg
30353381 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355310 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD365875100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 439134-46-0
Molekylformel C16H12Cl2
Molekylvikt (g/mol) 275.17
Synonym Naphthalene, 1-(3,4-dichlorophenyl)-1,2-dihydro- (9CI, ACI), 1-(3,4-Dichlorophenyl)-1,2-dihydronaphthalene (ACI)
LEDER Clc1ccc(cc1Cl)C2CC=Cc3ccccc23
Specifikationer
Kemiskt namn eller material 1-(3,4-Dichlorophenyl)-1,2-dihydronaphthalene
Kvantitet 100 mg
InChI formel InChI=1S/C16H12Cl2/c17-15-9-8-12(10-16(15)18)14-7-3-5-11-4-1-2-6-13(11)14/h1-6,8-10,14H,7H2
Formel vikt 274.0316