Läs mer
1-(3,4-Dihydroxyphenyl)-2-[(1-methylethyl)amino]-ethanone Hydrochloride, TRC
Produktkod. 30379295
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30379295 250 mg
1 options
Produktkod. 30379295 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD448195250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(3,4-dihydroxyphenyl)-2-[(1-methylethyl)amino]ethanone hydrochloride
CAS 16899-81-3
Molekylformel C11 H15 N O3 . HCl
Kvantitet 250 mg
Synonym Isoprenaline Imp. A (EP) as Hydrochloride, 1-(3,4-Dihydroxyphenyl)-2-[(1-methylethyl)amino]ethanone hydrochloride (1:1), 3',4'-Dihydroxy-2-(isopropylamino)acetophenone hydrochloride, 3,4-Dihydroxy-α-(isopropylamino)acetophenone hydrochloride, U 12844A
InChI formel InChI=1S/C11H15NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,12-14H,6H2,1-2H3;1H
LEDER Cl.O=C(C1=CC=C(O)C(O)=C1)CNC(C)C
IUPAC-namn 1-(3,4-dihydroxyphenyl)-2-(propan-2-ylamino)ethanone;hydrochloride
Molekylvikt (g/mol) 245.7
Formel vikt 245.0819