missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62681.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499168

  • 3427.64 SEK / 1g


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD06173487

Specifications

1-(3,4-Dimethylphenyl)-3-(4-fluorophenyl)urea
MFCD06173487
Solid
1g
258.29
97%
Videos
SDS