Läs mer
1-(3,4-Dimethylphenyl)-3-methyl-1H-pyrazol-5(4H)-one, TRC
Produktkod. 30180346
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180346 5 mg
30180138 25 mg
30180080 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD9485985MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18048-64-1
Molekylformel C12 H14 N2 O
Molekylvikt (g/mol) 202.25
Synonym 3H-Pyrazol-3-one, 2-(3,4-dimethylphenyl)-2,4-dihydro-5-methyl-, 2-Pyrazolin-5-one, 3-methyl-1-(3,4-xylyl)- (8CI), 2-(3,4-Dimethylphenyl)-2,4-dihydro-5-methyl-3H-pyrazol-3-one, 3-Methyl-1-(3,4-dimethylphenyl)-2-pyrazolin-5-one, SB-392033, Eltrombopag Stage 5
IUPAC-namn 2-(3,4-dimethylphenyl)-5-methyl-4H-pyrazol-3-one
LEDER CC1=NN(C(=O)C1)c2ccc(C)c(C)c2
Specifikationer
Kemiskt namn eller material 3-methyl-1-(3,4-xylyl)-2-Pyrazolin-5-one
Kvantitet 5 mg
InChI formel InChI=1S/C12H14N2O/c1-8-4-5-11(6-9(8)2)14-12(15)7-10(3)13-14/h4-6H,7H2,1-3H3
Formel vikt 202.1106