Läs mer
1-(3,4-Methylenedioxyphenyl)-4,4-dimethyl-pent-1-en-3-one, TRC
Produktkod. 30477470
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477470 100 mg
30464418 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477470 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(3,4-Methylenedioxy-phenyl)-4,4-dimethyl-pent-1-en-3-one
CAS 2419-68-3
Molekylformel C14 H16 O3
Kvantitet 100 mg
InChI formel InChI=1S/C14H16O3/c1-14(2,3)13(15)7-5-10-4-6-11-12(8-10)17-9-16-11/h4-8H,9H2,1-3H3/b7-5+
LEDER CC(C)(C)C(=O)\C=C\c1ccc2OCOc2c1
IUPAC-namn (E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-one
Molekylvikt (g/mol) 232.28
Formel vikt 232.1099