Läs mer
1,3,4-Oxadiazole, TRC
Produktkod. 30428312
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428312 100 mg
30414859 10 mg
30420299 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428312 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO990985100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 288-99-3
Molekylformel C2H2N2O
Molekylvikt (g/mol) 70.05
IUPAC-namn 1,3,4-oxadiazole
LEDER o1cnnc1
Specifikationer
Kemiskt namn eller material 1,3,4-Oxadiazole
Kvantitet 100 mg
InChI formel InChI=1S/C2H2N2O/c1-3-4-2-5-1/h1-2H
Formel vikt 70.0167