Läs mer
1-[3-(4-tert-Butylbenzoyl)propyl]-4-hydroxypiperidine, TRC
Produktkod. 30136650
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136650 5 g
17924615 1 g
17902705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6903905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97928-18-2
Molekylformel C19 H29 N O2
Molekylvikt (g/mol) 303.44
Synonym 1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one, Ebastine Imp. D (EP)
IUPAC-namn 1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
LEDER CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(O)CC2
Specifikationer
Kemiskt namn eller material 1-[4-(1,1-dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C19H29NO2/c1-19(2,3)16-8-6-15(7-9-16)18(22)5-4-12-20-13-10-17(21)11-14-20/h6-9,17,21H,4-5,10-14H2,1-3H3
Formel vikt 303.2198
Procent renhet >95%