Läs mer
1-(3,5-Di-tert-butyl-4-hydroxyphenyl)-1,2-ethanediol, TRC
Produktkod. 30365697
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365697 5 mg
30362072 25 mg
30356030 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365697 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1256505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125489-21-6
Molekylformel C16H26O3
Molekylvikt (g/mol) 266.38
Synonym 1-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]-1,2-ethanediol
IUPAC-namn 1-(3,5-ditert-butyl-4-hydroxyphenyl)ethane-1,2-diol
LEDER CC(C)(C)c1cc(cc(c1O)C(C)(C)C)C(O)CO
Specifikationer
Kemiskt namn eller material 1-(3,5-Di-tert-butyl-4-hydroxyphenyl)-1,2-ethanediol
Kvantitet 5 mg
InChI formel InChI=1S/C16H26O3/c1-15(2,3)11-7-10(13(18)9-17)8-12(14(11)19)16(4,5)6/h7-8,13,17-19H,9H2,1-6H3
Formel vikt 266.1882