Learn More
Alfa Aesar™ 1-(3,5-Dimethylphenyl)-3-(4-fluorophenyl)urea, 97%
Brand:  Alfa Aesar™ H62710.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15449198
  • 12447.17 SEK / 5g
Estimated Shipment date
from Supplier 14-11-2018
Add to basket
Specifications
Chemical Identifiers
890342-37-7
258.30
HRCNESVWHQIOFC-UHFFFAOYSA-N
16374815
C15H15FN2O
MFCD07091480
1-3,5-dimethylphenyl-3-4-fluorophenyl urea, 3-3,5-dimethylphenyl-1-4-fluorophenyl urea
Specifications
1-(3,5-Dimethylphenyl)-3-(4-fluorophenyl)urea
890342-37-7
MFCD07091480
HRCNESVWHQIOFC-UHFFFAOYSA-N
16374815
Solid
5g
C15H15FN2O
1-3,5-dimethylphenyl-3-4-fluorophenyl urea, 3-3,5-dimethylphenyl-1-4-fluorophenyl urea
258.30
258.29
97%