missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine-2,4,6-trithiol trisodium salt, approx. 15% solution in water, ACROS Organics™

355.04 SEK - 2398.42 SEK

Chemical Identifiers

CAS 17766-26-6
Molecular Formula C3N3Na3S3
Molecular Weight (g/mol) 243.22
InChI Key YQHCWFVOBRIHOM-UHFFFAOYSA-N
Synonym 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, trisodium salt, 1,3,5-triazine-2,4,6-trithiol trisodium salt, trithiocyanuric acid trisodium
PubChem CID 66735895
IUPAC Name sodium;1,3,5-triazinane-2,4,6-trithione
SMILES C1(=S)NC(=S)NC(=S)N1.[Na].[Na].[Na]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10431854
SDS View Documents
Acros Organics
426540010
1L Glass bottle 958.94 SEK
1 L View alternative products
Save Up to 
»
Estimated Shipment
04-09-2020
Add to basket
 
10714755
SDS View Documents
Acros Organics
426540025
2.5L Glass bottle 2398.42 SEK
2.50 L View alternative products
Save Up to 
»
Estimated Shipment
21-12-2020
Add to basket
 
10102774
SDS View Documents
Acros Organics
426542500
250mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications

Chemical Identifiers

17766-26-6
243.22
1,3,5-triazine-2,4,6 1h,3h,5h-trithione, trisodium salt, 1,3,5-triazine-2,4,6-trithiol trisodium salt, trithiocyanuric acid trisodium
sodium;1,3,5-triazinane-2,4,6-trithione
C3N3Na3S3
YQHCWFVOBRIHOM-UHFFFAOYSA-N
66735895
C1(=S)NC(=S)NC(=S)N1.[Na].[Na].[Na]

Specifications

1,3,5-Triazine-2,4,6-trithiol trisodium salt
Authentic
1.12
101°C
-3°C
17766-26-6
1,3,5-triazine-2,4,6 1h,3h,5h-trithione, trisodium salt, 1,3,5-triazine-2,4,6-trithiol trisodium salt, trithiocyanuric acid trisodium
YQHCWFVOBRIHOM-UHFFFAOYSA-N
sodium;1,3,5-triazinane-2,4,6-trithione
66735895
Liquid
1.1200g/mL
Glass bottle
1.6 mPa.s (20°C)
Colorless to Yellow
1L
C3N3Na3S3
Solubility in water: miscible
C1(=S)NC(=S)NC(=S)N1.[Na].[Na].[Na]
243.22
243.22
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3,5-Triazine-2,4,6-trithiol trisodium salt, approx. 15% solution in water

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.