Läs mer
1,3,5-Tribromo-2-(2-propen-1-yloxy)-benzene, TRC
Produktkod. 17943655
Change view
Kvantitet:
1 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943655 50 g
17921555 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17943655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77190050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3278-89-5
Molekylformel C9 H7 Br3 O
Molekylvikt (g/mol) 370.863
Synonym 2,4,6-Tribromophenyl allyl ether, Benzene, 1,3,5-tribromo-2-(2-propenyloxy)- (9CI), Ether, allyl 2,4,6-tribromophenyl (7CI,8CI), 1,3,5-Tribromo-2-(2-propen-1-yloxy)benzene, 1,3,5-Tribromo-2-prop-2-enoxybenzene, 2,4,6-Tribromophenyl allyl ether, 2-Allyloxy-1,3,5-tribromobenzene, Allyl 2,4,6-tribromophenyl ether, NSC 35767, Pyroguard FR 100, TBPAE
IUPAC-namn 1,3,5-tribromo-2-prop-2-enoxybenzene
LEDER Brc1cc(Br)c(OCC=C)c(Br)c1
Specifikationer
Kemiskt namn eller material 1,3,5-Tribromo-2-(2-propen-1-yloxy)-benzene
Kvantitet 50 g
InChI formel InChI=1S/C9H7Br3O/c1-2-3-13-9-7(11)4-6(10)5-8(9)12/h2,4-5H,1,3H2
Formel vikt 367.8047