missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-trichloro-2-iodobenzene, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | 2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene |
Formula Weight | 307.35 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3, 5-Trichloro-2-iodobenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'