Läs mer
1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, TRC
Produktkod. 30456888
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30456888 10 g
1 options
Produktkod. 30456888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79762510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Plastic additive 10
CAS 1709-70-2
Molekylformel C54 H78 O3
Kvantitet 10 g
Synonym 4,4',4''-[(2,4,6-Trimethyl-1,3,5-benzenetriyl)tris(methylene)]tris[2,6-bis(1,1-dimethylethyl)phenol, α,α',α''-(2,4,6-Trimethyl-s-phenenyl)tris[2,6-di-tert-butyl-p-cresol, α,α',α''-(Trimethyl-s-phenenyl)tris[2,6-di-tert-butyl-p-cresol, 1,3,5-Dimethyl-2,4,6-tris(3,5-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tri(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxytolyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(4-hydroxy-3,5-di-tert-butylbenzyl)benzene, 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-2,4,6-trimethylbenzene, 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxyphenylmethyl)-2,4,6-trimethylbenzene, 1,3,5-Trismethyl-2,4,6-tris(3,5-tert-butyl-4-hydroxybenzyl)benzene, 2,4,6-Bis(4-hydroxy-3,5-di-tert-butylbenzyl)mesitylene, 2,4,6-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)mesitylene, 2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene, 2,4,6-Tris(4-hydroxy-3,5-di-tert-butylbenzyl)-1,3,5-trimethylbenzene, 2,4,6-Tris(4-hydroxy-3,5-di-tert-butylbenzyl)mesitylene, 3,3',3'',5,5',5''-Hexa-tert-butyl-α,α',α''-(mesitylene-2,4,6-triyl)tri-p-cresol, ADK Stab A 330, ADK Stab AO 330, AO 330, AO 40, Agidol 40, Alvinox 100, Anox 330, Anox 330TDS, Antioxidant 1330, Antioxidant 330, Antioxidant 40, Ethanox 300, Ethanox 330, Ethyl 330, Ethyl Antioxidant 330, Good-rite 1330, IR 1330, Ionox 330, Irg 1330, Irganox 1330, Irganox 330, Mark AO 330, NSC 85846, Seenox 326M, Yoshinox 1330, Plastic additive 10
InChI formel InChI=1S/C54H78O3/c1-31-37(22-34-25-40(49(4,5)6)46(55)41(26-34)50(7,8)9)32(2)39(24-36-29-44(53(16,17)18)48(57)45(30-36)54(19,20)21)33(3)38(31)23-35-27-42(51(10,11)12)47(56)43(28-35)52(13,14)15/h25-30,55-57H,22-24H2,1-21H3
LEDER Cc1c(Cc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)c(C)c(Cc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)c(C)c1Cc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C
IUPAC-namn 4-[[3,5-bis[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]-2,4,6-trimethylphenyl]methyl]-2,6-ditert-butylphenol
Molekylvikt (g/mol) 775.2
Formel vikt 774.5951